missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bio-Serv™ Prima-Treats™ Animal Diet
Healthy treat used for training or positive reward following a less desirable procedure.
$274.32 - $294.09
Specifications
Product Type | Animal Diet |
---|---|
Components | Dextrose, Casein, Sucrose, Cellulose, Mineral Mix, Canola Oil, Natural and Artificial Flavors, Magnesium Silicate, Choline Dihydrogen Citrate, Vitamin Mix, Calcium Silicate, Ascorbic Acid, DL-Methionine, Magnesium Stearate, Food Dye, BHA |
Particle Shape | Wafer |
Quantity | 1000/Box |
Diameter (English) | 1 in. |
Catalog Number | Mfr. No. | Quantity | Flavor | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Flavor | Price | Quantity | ||||
14-726-454
|
Bio-Serv™
F0295 |
1000/Box | Apple |
Each for $294.09
|
|
||||
14-726-456
|
Bio-Serv™
F0300 |
1000/Box | Banana |
Each for $294.09
|
|
||||
14-726-460
|
Bio-Serv™
F0308 |
1000/Box | Orange |
Each for $294.09
|
|
||||
14726698
|
Bio-Serv™
F0293 |
1000/Box | Blueberry |
Each for $274.32
|
|
||||
Description
- Comes in variety of flavors
- Nutritionally complete treat that compliments enrichment programs
- Less labor intensive and more sanitary than perishable fruit
- Excellent source of vitamin C
- Nutritionally assayed
- Weight: 5g (0.18 oz.)
- Shape: Wafer
- Packaging:250/Jar and 1000/Box.
Specifications
Animal Diet | |
Wafer | |
1 in. |
Dextrose, Casein, Sucrose, Cellulose, Mineral Mix, Canola Oil, Natural and Artificial Flavors, Magnesium Silicate, Choline Dihydrogen Citrate, Vitamin Mix, Calcium Silicate, Ascorbic Acid, DL-Methionine, Magnesium Stearate, Food Dye, BHA | |
1000/Box | |
2.54 cm |