Epredia™ PrintMate™ AS Cassette Printer Accessories
Maintain the Epredia™ PrintMate™ Cassette Printer with PrintMate™ Accessories.
Supplier:  Epredia™ A79510013
 View more versions of this product
Catalog No. A79510013
Explore available promotions
Description
Description
Accessories Include:
  • Collection system collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
Specifications
Specifications
Hot Foil Tape
Hot Foil Tape, package of 5 rolls, 5 cardboard tubes, one leader
PrintMate AS Cassette Printer, Printmate Cassette Printer