Epredia™ PrintMate™ AS Cassette Printer Accessories
Maintain the Epredia™ PrintMate™ Cassette Printer with PrintMate™ Accessories.
Supplier: Epredia™ A79510024
Description
Accessories Include:
- Collection system collection trays (box of 4)
- Hopper
- Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
Specifications
Cassette Hopper | |
Cassette Hopper (each) | |
PrintMate AS Cassette Printer, PrintMate Cassette Printer |