missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Pro-Detect™ Rapid GST Assay Kit
Pro-Detect™ Rapid Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of tagged proteins in protein expression and purification systems
Supplier: Thermo Scientific™ A38502
Includes: 10 assays
Description
- Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the tag to provide a visual, color readout for detection within ten minutes.
- Detect proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling.
- Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
- Dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
- Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
- Specific—assay does not cross-react with common lysis and protein preparation reagents
- Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
- Long shelf life—stable for at least one year at 4°C
Specifications
• 10 Rapid Assay Strips • Sample Diluent, 15 mL Sufficient for 10 assay determinations Store at 2–8°C. |
|
Lateral Flow Rapid Assay | |
Pro-Detect™ | |
Protein tag | |
Lateral Flow Rapid Assay | |
Wet Ice |
GST-tagged proteins | |
Chromogenic | |
Assay | |
10 Assays | |
10 Assays | |
Protein |
For Research Use Only. Not for use in diagnostic procedures.