missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pro-Lab Diagnostics™ Prolex™ - Blue E. coli 0157 Latex Test Reagent Kits
Just one colony is sufficient for the test, enabling even limited growth to be detected
Supplier: Pro-Lab Diagnostics™ PL070B
Description
- Developed to remove the possibility of cross-reactions with E. hermanii and other sorbitol negative Escherichia species
- Latex agglutination
- Test results in 2 minutes
- CLIA Complexity: Non-waived
- Sensitivity of 100%
Specifications
2°C to 8°C | |
E. coli serogroup O157 | |
Blue E. coli O157 Latex Test Reagent Kit | |
50 Tests |
Non-waived | |
1 | |
E. coli O157 Latex reagent 3.1mL, negative control Latex reagent 1.5mL, positive control 1.5mL, Agglutination cards 25, mixing sticks 75 | |
2 min. |