Learn More
Thermo Scientific Chemicals Procaine hydrochloride, 99%
Supplier: Thermo Scientific Chemicals 207311000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Procaine hydrochloride | |
154.0°C to 158.0°C | |
5 to 6.5 (2% w/v aq. soln.) | |
Authentic | |
Plastic bottle | |
H2NC6H4CO2CH2CH2N(C2H5)2·HCl | |
100 g | |
15, 7875 | |
Solubility in water: soluble. Other solubilities: soluble in chloroform, practically insoluble in ether | |
[H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1 | |
272.77 | |
CHEBI:8431 | |
99% |
51-05-8 | |
Colorless to White | |
0.5% max. (1 g, 105°C) | |
99% | |
C13H21ClN2O2 | |
MFCD00013000 | |
14, 424 | |
procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor | |
HCBIBCJNVBAKAB-UHFFFAOYSA-N | |
2-(diethylamino)ethyl 4-aminobenzoate;hydrochloride | |
5795 | |
272.77 | |
Crystalline Powder or Crystals |
Chemical Identifiers
51-05-8 | |
272.77 | |
HCBIBCJNVBAKAB-UHFFFAOYSA-N | |
5795 | |
2-(diethylamino)ethyl 4-aminobenzoate;hydrochloride |
C13H21ClN2O2 | |
MFCD00013000 | |
procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor | |
CHEBI:8431 | |
[H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 200-077-2
RUO – Research Use Only