missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gaylord Chemical Company, LLC PROCIPIENT™ (Dimethyl Sulfoxide USP, PhEur)

Procipient™ DMSO is both an active pharmaceutical ingredient and a versatile excipient that is widely used in various pharmaceutical and biotechnology applications. Procipient is an extremely polar, water-soluble solvent with a minimum purity of 99.99%.

Supplier:  Gaylord Chemical Company, LLC 5010430LT

 View more versions of this product

Catalog No. 18-614-918


Explore available promotions

Description

Description

Procipient™ Characteristics:

  • Highly purified DMSO with a minimum purity of 99.99%
  • Water miscible/extremely polar solvent
  • Odorless, clear water-white liquid
  • Essentially non-toxic by all routes of administration
  • Chemically compatible with other excipients
  • Low viscosity (2.0 cP at 25°C)
  • Density similar to water
  • (1.0955 g / mL at 25°C)
  • Low volatility: 189°C boiling point
Specifications

Specifications

Dimethyl Sulfoxide
67-68-5
1.0955 g/mL (25°C)
1 L
215°C (419°F)
Liquid
59.4 Pa at 20°C (68°F)
Water Miscible, Extremely Polar Solvent
CS(C)=O
78.13
USP
18.5°C (65.3°F)
Clear
189°C
87°C (188.6°F) (Closed Cup)
Odorless
C2H6OS
2.0 cP (25°C)
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methanesulfinylmethane
99.99%

Chemical Identifiers

67-68-5
78.13
methanesulfinylmethane
C2H6OS
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
CS(C)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store in a well-ventilated place. Keep cool.

SDS
Documents

Documents

Promotions

Promotions