Bel-Art™ ProCulture™ Cordless Homogenizer Unit
The ProCulture™ Cordless Homogenizer Unit is a hand-held homogenizer and is ideal for thorough and faster processing of samples.
Supplier:  Bel-Art™ F651000000
Catalog No. 17000201
Explore available promotions
Description
Description
  • Hand-held, cordless homogenizer features a simple push-in, pull-out pestle exchange and is ideal for thorough and faster processing of one-to-many samples
  • Optional ProCulture™ Matched Pestles and Tubes assure tight tolerances and thorough homogenization and grinding of materials resulting in optimal efficiency of pellet reconstitution or re-suspension; F65000-0005, F65000-0006, F65000-9006
  • Optional ProCulture™ Pestles have a unique ribbed shaft that provides precise finger control of homogenization for shear sensitive samples; also fit most motorized mixer adapters; F65000-0001, F65000-0002, F65000-9001, F65000-9002
Specifications
Specifications
Cordless Homogenizer Unit
Yes
Yes
CE
Homogenizer with Cordless Motor, AA Batteries (2), and Pestle Adapter
Videos
SDS
Documents
Documents
Promotions
Promotions
WARNING: This product can expose you to chemicals which are known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.