missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ ProCulture Warming Plate, Ambient to 37°C, Aluminum
Shape: Rectangle; Size: 7 x 8 in.
$1010.93 - $1010.93
Specifications
Shape | Rectangle |
---|---|
Type | Warming Plate |
Top Plate Material | Aluminum |
Size | 7 x 8 in. |
Temperature (Metric) | Ambient to 37°C |
Catalog Number | Mfr. No. | Voltage | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Voltage | Price | Quantity | |||||
14-513-613
|
Bel-Art™
F370150000 |
120 V |
Each for $1,010.93
|
|
|||||
Description
Shape: Rectangle; Size: 7 x 8 in.
- Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37°C (±1°C) outside of the incubator.
- Avoids numerous entries into the CO2 incubator which alter its atmosphere, harming ongoing experiments in the incubated cultures
- Reduces cell stress and allows enzymes like trypsin to work efficiently
- Can be used to maintain the warmth of reagents inside the hood
- Fits easily in cell culture hoods or on the workbench
- Temperature remains uniform across the entire anodized aluminum warming plate; measures 17.8 D x 20.3 cmW (7 x 8 in.)
- 15-minute count down timer keeps track of time sensitive applications
- UV resistant polypropylene housing will not break down under germicidal UV lights
- Water resistant; safe for cleaning with Ethanol or IPA
- 220V is CE marked
Specifications
Rectangle | |
Aluminum | |
Ambient to 37°C | |
25.4 cm | |
9.75 in. | |
24.9 cm |
Warming Plate | |
7 x 8 in. | |
10 in. | |
2.25 in. | |
5.7 cm | |
22 w |