missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Productainer™ BioProcess Container (BPC), 200 L with 2 Top and 1 Bottom Ports, 3D Single-Use

Using these BPC bags with 2 top and 1 bottom ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Supplier:  Thermo Scientific™ SH3065003

Catalog No. SH3065003


Explore available promotions

Description

Description

Using Thermo Scientific™ Productainer™ BPC bag with 2 top and 1 bottom ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Specifications

200 L
Packages placed in strong cardboard cartons with product and lot identification
Each
Gamma Irradiated
Line 3. MPX Insert, polycarbonate Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 12.7 x 19.1 mm (0.5 in. x 0.75 in.) Bottom Port
Productainer™
Bottom Port: 1x 0.5 in., Top Ports: 1 x 0.25 in. 1 x 0.5 in.
Polyethylene
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
SV50517.06
Line 1. MPX Body, polycarbonate, Tubing: C-Flex, length: 45.7 cm (18 in.) ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) Top Port
Line 2. Body luer lock connection, polypropylene Tubing: C-Flex, length: 30.5 cm (12 in.) ID x OD: 6.4 x 9.5mm (0.25 x 0.38 in.) Top Port
Fewer resources used and less waste
CX5-14
Videos
SDS
Documents

Documents

Promotions

Promotions