missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Productainer™ BioProcess Container (BPC), 200L with 4 Top Drains, 3D Single-Use BPC

Using these BPC bags with 4 top drains, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Supplier:  Thermo Scientific™ SH3065303

Catalog No. SH3065303


Explore available promotions

Description

Description

Using Thermo Scientific™ Productainer™ BPC bag with 4 top drains, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Specifications

200 L
200 L dip tube: 81 cm (32 in.)
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Gamma Irradiated
Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
Line 4. MPX body, polycarbonate Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 12.7 x 19.1 mm (0.5 in. x 0.75 in.) dip tube length: 30.5 cm (12 in.)
Fewer resources used and less waste
CX5-14
Polyethylene
Packages placed in strong cardboard cartons with product and lot identification
SV50076.04
Line 1. MPX insert, polycarbonate, Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) with dip tube length dependent on size
Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
Productainer™
Top Ports: 2 x 0.25 in. 2 x 0.5 in.
Videos
SDS
Documents

Documents

Promotions

Promotions