missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 816 Thermal Fingerless Winter Work Gloves/Flip-Top Mittens

Mittens feature a soft microfleece lining for cold-weather protection and easily convert to half-finger gloves for detail work.

$56.82 - $56.82

Specifications

Color Black
Hand Specific
Material Polyester
EN388 Standard 3423 X
For Use With (Application) Construction, Cold Weather Detail Work, Cold Storage/Refrigeration, and Outdoor Recreation
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-820-157
View Documents
Ergodyne™
17342
Small
Pair for $56.82
Only null left
 
19-820-158
View Documents
Ergodyne™
17343
Medium
Pair for $56.82
Only null left
 
19-820-160
View Documents
Ergodyne™
ERG17345
X-Large
Pair for $56.82
Only null left
 
19-820-161
View Documents
Ergodyne™
17346
2X-Large
Pair for $56.82
Only null left
 
Description

Description

  • Mittens convert to half-finger gloves for detailed work
  • Flip-top mitten and thumb stow easily in mitten pocket when not in use
  • Soft microfleece lining provides comfort, flexibility, and protection in cold conditions
  • Rugged synthetic leather palm maintains flexibility in cold temps
  • Reinforced thumb saddle extends glove life
  • Contoured gathered elastic cuff with ID space
  • Gloves fit over jacket sleeves for convenient on/off wear
  • Reflective accents enhance wearer visibility and hand signaling
  • Soft thumb nose/brow wipe area
Specifications

Specifications

Black
Polyester
Construction, Cold Weather Detail Work, Cold Storage/Refrigeration, and Outdoor Recreation
Microfleece
ProFlex™ 816
Specific
3423 X
Elasticated
Synthetic Leather
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'