Sartorius Proline™ Plus Single-Channel Mechanical Pipetter
Designed to offer comfort and quality for everyday manual pipetting. Sartorius Proline™ Plus Single-Channel Mechanical Pipetter combines durable construction with ease of use, plus the widest range of pipettes for various users and applications. Available with volume ranges from 0.1μL to 10mL.
Supplier:  Sartorius 728020
 View more versions of this product
Catalog No. 14-559-428
Explore available promotions
Description
Description
Ideal for experienced laboratory professionals and students alike
  • Widest range of pipettes, including fixed volume pipettes for when volumes need to be ready-set to avoid errors
  • Optiload mechanism allows tip loading with perfect tip sealing on the tip cone
  • Safe-Cone Filters minimize risk of contamination
  • Adjustable volume setting with click stop
  • Thermal insulation of internal components improves accuracy
  • Durable, strengthened structure with high chemical and UV resistance
User-friendly and ergonomic design
  • Light pipetting forces, comfortable handle and finger support
  • No need to squeeze the pipetter during pipetting
  • Light and easy tip ejection
  • Reduces the risk of repetitive stress injury
  • Easy to use, do-it-yourself calibration
  • Large numbers and color-coding on the volume display
  • Volume color-coding enables correct tip volume selection through matching color code
  • Fully autoclavable without disassembling
  • Easy to clean, with only three parts to disassemble
Specifications
Specifications
5 to 1%
Autoclavable
Minimised Risk of Contamination with Safe Cone Filters and Full Autoclavability
Proline™
0.5 to 10 μL
Minimised risk of contamination with safe cone filters and full autoclavability
4.00 to 0.60
2 Years
ISO 9001/13485, ISO 14001
Gray
1
Pipette
Ergonomic
0.01μL
Yes
SDS
Documents
Documents
Accessories