missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sartorius Proline™ Plus Single-Channel Mechanical Pipetter
Designed to offer comfort and quality for everyday manual pipetting. Sartorius Proline™ Plus Single-Channel Mechanical Pipetter combines durable construction with ease of use, plus the widest range of pipettes for various users and applications. Available with volume ranges from 0.1μL to 10mL.
Supplier: Sartorius 728060
Description
Ideal for experienced laboratory professionals and students alike- Widest range of pipettes, including fixed volume pipettes for when volumes need to be ready-set to avoid errors
- Optiload mechanism allows tip loading with perfect tip sealing on the tip cone
- Safe-Cone Filters minimize risk of contamination
- Adjustable volume setting with click stop
- Thermal insulation of internal components improves accuracy
- Durable, strengthened structure with high chemical and UV resistance
- Light pipetting forces, comfortable handle and finger support
- No need to squeeze the pipetter during pipetting
- Light and easy tip ejection
- Reduces the risk of repetitive stress injury
- Easy to use, do-it-yourself calibration
- Large numbers and color-coding on the volume display
- Volume color-coding enables correct tip volume selection through matching color code
- Fully autoclavable without disassembling
- Easy to clean, with only three parts to disassemble
Specifications
2 to 0.6% | |
Autoclavable | |
Minimised Risk of Contamination with Safe Cone Filters and Full Autoclavability | |
Proline™ | |
20 to 200 μL | |
Minimised risk of contamination with safe cone filters and full autoclavability | |
0.80 to 0.15 | |
2 Years |
ISO 9001/13485, ISO 14001 | |
Yellow | |
1 | |
Pipette | |
Ergonomic | |
0.2μL | |
Yes |