missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega 100bp DNA Ladder Molecular Weight Marker DFS Item

11 fragments from 100bp to 1,000bp in 100bp increments and includes an additional 1.5kb band

Supplier:  Promega G2101

Catalog No. PR-G2101


Explore available promotions

Includes: 6X Blue/Orange Loading Dye

Description

Description

  • Eleven Fragments: 100-1,000bp in 100bp Increments Plus a Band at 1,500bp
  • Load 5μL/lane
  • Supplied at a concentration of 0.13μg/μL
  • Supplied with 6X Blue/Orange Loading Dye
Specifications

Specifications

Blue, Orange
DNA Molecular Weight Marker/Ladder
Determining the size of DNA fragment
11
−30 to −10°C
0.13 μg/μL
Liquid
1 × 250μL 100bp DNA ladder, 1 × 1mL blue/orange 6X loading dye
250 μL
Videos
SDS
Documents

Documents

Promotions

Promotions