Promega 50bp DNA Step Ladder Molecular Weight Marker for Electrophoresis DFS Item
Sixteen DNA fragments ranging from 50bp to 800bp in 50bp increments plus an 1,800bp backbone fragment.
Supplier:  Promega G4521
Catalog No. PR-G4521
Explore available promotions
Includes: 6X Blue/Orange Loading Dye
Description
Description
  • Sixteen DNA Fragments of 50-800bp in 50bp Increments
  • Includes an 1,800bp backbone fragment
  • Load 5μL/lane
  • Supplied at a concentration of 0.34μg/μL
  • Supplied with 6X Blue/Orange Loading Dye
Specifications
Specifications
10mM Tris HCl pH 7.5, 1mM EDTA
0.34 μg/μL
Liquid
Separation, size determination and quantitation of DNA and RNA fragments
90 μg
Blue, Orange
DNA Molecular Weight Marker/Ladder
90μg 50bp DNA Step ladder, 1mL blue/orange 6X loading dye
16
−30 to −10°C
Product Suggestions
Product Suggestions