Promega ADP-Glo™ Max Assay
The ADP-Glo™ Max Assay is a luminescent ADP detection assay that provides a universal, homogeneous, high-throughput screening method to measure ATPase or kinase activity by quantifying the amount of ADP produced in a reaction.
Supplier: Promega V7001
Description
- Luminescent Method to Measure ATPase or Kinase Activity
- Screen library compounds for effects on target enzymes
- Perform mode of action studies on enzymes of interest
- Measure activity of immunoprecipitated ATPases/kinases
- Use when higher ATP concentration is required (up to 5mM)
Specifications
ADP-Glo Max Assay | |
Screen library compound for effect on target Enzyme, perform mode of action studies on Enzyme of interest, measure activity of immunoprecipitated ATPases/Kinase | |
1000 Assays |
1000 Assay | |
1 vial ADP-Glo Max detection substrate (lyophilized), 10 mL ADP-Glo Max detection buffer, 2 x 500 μL ADP, 10mM, 250 μL Ultra pure ATP, 100mM, 5 mL ADP-Glo reagent | |
-30°C to -10°C |