Promega BenchTop DNA Markers DFS Item
Ready-to-load markers offer the convenience of room temperature storage
Supplier:  Promega G7531
 View more versions of this product
Catalog No. PR-G7531
Explore available promotions
Description
Description
  • Six Bands of Equal Intensity: 50, 150, 300, 500, 750, and 1,000bp
  • Store at room temperature (22-25°C)
  • Premixed with loading buffer; ready to load onto agarose gels
  • Load 6μL/lane for agarose gels
Specifications
Specifications
Blue, Orange
DNA Molecular Weight Marker/Ladder
Supplied in a stabilizing solution of 1X blue/orange loading dye
6
−30 to +30°C
6.1 ng/μL
Liquid
Separation, size determination and quantitation of DNA and RNA fragments
300 μL