Promega BenchTop DNA Markers
Ready-to-load markers offer the convenience of room temperature storage
Supplier: Promega G8291
Description
- Eleven Fragments of 100-1,000bp in 100bp Increments Plus a Band at 1,500bp
- Store at room temperature (22-25°C)
- Premixed with loading buffer; ready to load onto agarose gels
- Load 6μL/lane
Specifications
Blue, Orange | |
DNA Molecular Weight Marker/Ladder | |
Supplied in a stabilizing solution of 1X blue/orange loading dye | |
11 | |
−30 to +30°C |
108 ng/μL | |
Liquid | |
Separation, size determination and quantitation of DNA and RNA fragments | |
300 μL |