Learn More
Promega Blue/Orange Loading Dye, 6X DFS Item
Convenient marker dye used for loading DNA samples into gel electrophoresis wells and tracking migration during electrophoresis.
Supplier:  Promega G1881
Numéro de catalogue. PR-G1881
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Quality tested and certified free of nuclease activity
Certifications
Certified to be free of nuclease activity.
Spécifications
Spécifications
Blue, Orange
DNA Stain
Used for loading DNA samples into Gel electrophoresis wells and tracking migration during electrophoresis
−30 to +10°C
6X
0.03% Bromophenol Blue, 0.03% Xylene Cyanol FF, 0.4% Orange G, 15% Ficoll™ 400, 10mM Tris HCl (pH 7.5), 50mM EDTA (pH 8)
3 x 1 mL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS