Promega Broad Range Protein Molecular Weight Marker DFS Item
10-225kDa; 9 bands; ready to load. Protein staining required for detection.
Supplier:  Promega V8491
Catalog No. PR-V8491
Explore available promotions
Description
Description
  • Nine Evenly Spaced Bands of 10-225kDa for Use with SDS-PAGE
  • 50kDa band is 3X intensity for use as a reference
  • Ready-to-load markers
  • Supplied at a concentration of 5μL/lane
Specifications
Specifications
5 μL/Lane
Designed for use with SDS electrophoresis
10 to 225 kd
100 Lanes
Protein Molecular Weight Marker/Ladder
100 Lanes
−30 to +10°C