Promega Calf Intestinal Alkaline Phosphatase (CIAP) Buffer Pack DFS Item
A complete system for phosphorylating both double- and single-stranded DNA and RNA with T4 PNK and [gamma-32P]ATP.
Supplier:  Promega M1833
Catalog No. PR-M1833
Explore available promotions
Description
Description
  • Phosphorylates RNA, ssDNA and dsDNA Ends with a Purchased Radiolabeled Nucleotide
  • System includes enzymes, buffers and control DNA standards for measuring reaction efficiencies (except radionucleotides)
  • Flexible labeling works with [γ-32P]ATP, [γ-33P]ATP or [γ-35S]ATP
Specifications
Specifications
Colorless
9.3
-30°C to -10°C
CIAP Buffer Pack
0.5M Tris HCl (pH 9.3 at 25°C), 10mM MgCl2, 1mM ZnCl2, 10mM Spermidine
Used with Alkaline Phosphatase Enzyme to preventing religation of linearized cloning vehicle DNA by removing Phosphate groups from both 5' terminals
3 x 500 μL
Liquid