Promega Caspase-Glo™ 8 and Caspase-Glo™ 9 Assay Systems DFS Item
Caspase-Glo™ Assay is a homogeneous luminescent assay that measures caspase-8 or caspase-9 activity.
Supplier:  Promega G8200
 View more versions of this product
Catalog No. PR-G8200
Explore available promotions
Includes: Caspase-Glo 8 Buffer or Caspase-Glo 9 Buffer; Caspase-Glo 8 Substrate or Caspase-Glo 9 Substrate
Description
Description
  • A Luminescent Assay That Measures Caspase-8 or 9 Activity
  • Decreased assay time and less enzyme needed
  • Homogeneous add-mix-measure protocol amenable to automation
  • U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
2.5 mL
0.5 to 1 hr.
Caspase-8 Activation Determination (EC50), Detection of Caspase-8 Activity, Caspase-8 Inhibition Determination (IC50)
1 x 1 Bottle Caspase-Glo™ 8 substrate, 1 x 2.5mL Caspase-Glo™ 8 Buffer, 1 x 7.5μL MG-132 inhibitor
-30°C to -10°C