Promega Caspase-Glo™ 8 and Caspase-Glo™ 9 Assay Systems
Caspase-Glo™ Assay is a homogeneous luminescent assay that measures caspase-8 or caspase-9 activity.
Supplier: Promega G8210
Includes: Caspase-Glo 8 Buffer or Caspase-Glo 9 Buffer; Caspase-Glo 8 Substrate or Caspase-Glo 9 Substrate
Description
- A Luminescent Assay That Measures Caspase-8 or 9 Activity
- Decreased assay time and less enzyme needed
- Homogeneous add-mix-measure protocol amenable to automation
- U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
2.5 mL | |
0.5 to 1 hr. | |
Caspase-9 Activation Determination (EC50), Caspase-9 Activity Determination, Caspase-9 Inhibition Determination (IC50) |
1 x 1 Bottle Caspase-Glo™ 9 substrate, 1 x 2.5mL Caspase-Glo™ 9 Buffer, 1 x 7.5μL MG-132 inhibitor | |
-30°C to -10°C |