missing translation for 'onlineSavingsMsg'
Learn More

Promega CellTiter 96™ Nonradioactive Cell Proliferation Assay (MTT) DFS Item

Colorimetric cell viability assay (MTT). Solubilization step is required.

$670.65 - $2073.91

Specifications

For Use With (Application) Cytotoxicity
Format 96-well Plates
Content And Storage COMPONENTS AT DIFFERENT TEMPS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR-G4000 DFS Item
View Documents
Promega
G4000
1000 Assays
Each for $670.65
Only null left
 
PR-G4100 DFS Item
View Documents
Promega
G4100
5000 Assays
Each for $2,073.91
Only null left
 
Description

Description

  • A Colorimetric Assay to Assess Cell Viability
  • Greater sensitivity than the neutral red procedure
  • Non-radioactive, fast and convenient assay applicable to a variety of cells
Specifications

Specifications

Cytotoxicity
96-well Plates
COMPONENTS AT DIFFERENT TEMPS
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'