Promega CytoTox 96™ Nonradioactive Cytotoxicity Assay
Colorimetric assay for the determination of cell membrane integrity based on detection of LDH release, an indicator of cytotoxicity. Sample transfer steps are required.
Supplier: Promega G1780
Description
- A Colorimetric Alternative to Radioactive Cytotoxicity Assays
- Measures release of LDH, an indicator of cytotoxicity
- Non-radioactive, time-saving assay
Specifications
Colorimetric | |
1000 Assays | |
COMPONENTS AT DIFFERENT TEMPS |
Total Cell Number Determination | |
10 x 96-well Plates |