Promega DeadEnd™ Fluorometric TUNEL System DFS Item
DNA fragmentation detection assay. Designed for the detection and quantitation of apoptotic cells within a cell population.
Supplier:  Promega G3250
Catalog No. PR-G3250
Explore available promotions
Includes: Plastic coverslips for simplified sample handling
Description
Description
  • A Classic TUNEL Assay to Detect and Quantitate Apoptotic Cells
  • Save time and money
  • Detect apoptosis in cultured cells or FFPE tissue sections
Specifications
Specifications
60 Reactions
1 × 60 Pack Plastic Coverslips, 1 × 9.6mL Equilibration Buffer, 6 × 50μL Nucleotide Mix, 1 × 70mL 20X SSC, 3 × 600U Terminal Deoxynucleotidyl Transferase (recombinant), 1 × 10mg Proteinase K
Detects and quantifies apoptotic cells within a cell population
6.8
-30°C to -10°C
Liquid