Learn More
Promega dNTP Mix DFS Item
A premixed solution containing sodium salts of dATP, dCTP, dGTP and dTTP, each at 10mM in water.
Supplier:  Promega U1515
 View more versions of this product
Catalog No. PR-U1515
Only null left
Explore available promotions
Description
Description
  • Ready to Use Solution of Deoxynucleotide Triphosphates
  • Highly pure (>99% triphosphate)
  • 10mM each nucleotide in water
  • Reduce pipetting steps by adding nucleotides as a mix
Specifications
Specifications
7.5
10 mM
1000 μL
-30°C to -10°C
>99%
PCR amplification
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.