FuGENE™ HD Transfection Reagent
FuGENE HD is a novel, nonliposomal reagent for transfecting DNA into a variety of cell lines with high efficiency and low toxicity.
Supplier: FuGENE™ E2311
Description
- Better Transfection for Challenging Cell Lines
- Difficult-to-transfect cell lines
- Transfection of primary cells
- Protein and virus production
Specifications
2°C to 10°C | |
Transfect DNA into a wide variety of cell lines with high efficiency and low toxicity, for use with luciferase assays, providing more expression and sensitive results | |
1 mL |
FuGENE™ HD is sold with a Limited Use Label License (LULL). You shall use this Product in accordance with all applicable laws, rules and regulations. You may not attempt to reverse engineer this Product. If you are not willing to accept these conditions, the distributor from whom the Product was purchased will accept the return of the unopened Product and provide you with a full refund. However, in the event this Product is opened, you agree to be bound by the conditions of this limited use statement.