Learn More
Promega GloMax™ 96 Microplate Luminometer Light Plate DFS Item
Quickly and easily verifies the performance of the GloMax 96 Microplate Luminometer
Supplier:  Promega E6531
Catalog No. PRE6531
Only null left
Explore available promotions
Description
Description
  • Checks sensitivity, reproducibility, and linearity
  • Consists of three highly stable light sources at signal levels spanning four decades
  • Powered by an easily replaceable battery
Specifications
Specifications
Light Plate
Verifies performance of GloMax 96 Microplate Luminometer
GloMax 96 Microplate Luminometer
Three highly stable light sources spanning four decades
Product Suggestions
Product Suggestions