Promega GoScript™ Reverse Transcription System
GoScript Reverse Transcriptase utilizes M-MLV and state-of-the-art buffer technology designed for qPCR to deliver robust, reliable cDNA synthesis of a full range of rare and abundant transcripts, even in the presence of inhibitors.
Supplier: Promega A5001
Includes: GoScript Reverse Transcriptase, GoScript 5X Reaction Buffer, MgCl2, Oligo(dT)15 Primer, PCR Nucleotide Mix, Random Primers, Recombinant RNasin Ribonuclease Inhibitor, Nuclease-free water
Description
- Reliable cDNA Synthesis for Sensitive Detection of Expression
- Available as a standalone enzyme, a complete reverse transcription kit or as a master mix with Oligo(dT) or Random Primers. RNasin(R) RNase Inhibitor protection included with Kit and Mixes
- Sensitive transcription of both high-copy and low-copy messages
- Transcribes short and long transcripts; processes through secondary structure
Specifications
• MgCl2, 1 x 1.2 mL • GoScript™ 5X Reaction Buffer, 1 x 600 μL • GoScript™ Reverse Transcriptase, 1 x 100 μL • oligo(dT)15 primer, 1 x 50 μg • Nuclease-free Water, 1 x 200μL Store at –30° to –10°C. |
|
RT-PCR, Real Time PCR (qPCR) | |
100 Reactions |
- U.S. Patent 6,242,235, Australian Patent 761757, Canadian Patent 2,335,153 and other patents and patents pending.
- U.S. Patent 5,552,302, Australian Patent 646803 and other patents.