Promega GoTaq™ Hot Start Polymerase: 5X DFS Item
GoTaq™ Hot Start Polymerase contains the high-performance GoTaq™ DNA Polymerase bound to a proprietary antibody that blocks polymerase activity until the reaction is heated at 94-95°C for two minutes.
Supplier:  Promega M5006
 View more versions of this product
Catalog No. PRM5006
Explore available promotions
Includes: GoTaq Hot Start Polymerase is supplied with 5X Colorless GoTaq Flexi Buffer, 5X Green GoTaq Flexi Buffer, and 25mM MgCl2.
Description
Description
  • Robust PCR with Convenient Room-Temperature Setup
  • Minimize nonspecific amplification and primer-dimers
  • Optimize reaction conditions with supplied Flexi Reaction Buffer and MgCl2 solution
  • Direct-to-gel convenience
  • Supplied at a concentration of 5u/μL
  • U.S. Patent No. 6,242,235, Australian Patent No. 761757, Canadian Patent No. 2,335,153.
  • Licensed under U.S. Patent Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.
Specifications
Specifications
5X Hot start Polymerase (5 x 500U), colorless flexi Buffer (20 x 1mL), 5X green flexi Buffer (20 x 1mL), MgCl2 (15 x 1.2mL). Store at −30 to −10°C
Cloning
5U/μL
2500U
2 min.