Promega GoTaq™ Long PCR Master Mix DFS Item
GoTaq Long PCR Master Mix contains the high-performance GoTaq Hot Start Polymerase in a specially formulated mixture with a proprietary thermal stable proofreading polymerase.
Supplier:  Promega M4021
Catalog No. PRM4021
Explore available promotions
Description
Description
  • Efficiently Amplify Long DNA Amplicons
  • Optimized blend of thermostable DNA polymerases with enhanced processivity and proofreading
  • Hot-start master mix for convenient room temperature set-up
  • Optimized components for enhanced yield, sensitivity and specificity
Specifications
Specifications
2X PCR master mix (2 x 1.25mL), Nuclease free Water (1 x 13mL), long PCR control primer pair (1 x 50μL), human genomic DNA (1 x 30μL). Store at −30 to −10°C
100
Real Time PCR (qPCR)
Long PCR master mix
100 Reactions