Promega GoTaq™ PCR Core Systems DFS Item
Includes GoTaq™ DNA polymerase and PCR Nucleotide Mix, along with high-performance buffers and magnesium chloride. Core System II also includes a Positive Control Plasmid DNA and Positive Control Primers. Performance-tested in PCR.
Supplier:  Promega M7660
Catalog No. PR-M7660
Description
Description
  • Convenient Bundles Containing Taq Polymerase and Reagents for PCR
  • PCR Core System I includes GoTaq(R), Mg-free Buffers, MgCl2, and PCR Nucleotide Mix
  • PCR Core System II also includes control primers, template
  • Direct-to-gel convenience with Green GoTaq(R) Flexi Buffer
Specifications
Specifications
1 x 1.2mL MgCl2, 1 x 200μL PCR nucleotide mix, 1 x 250μL GoTaq™ DNA Polymerase, 2 x 1mL 5X colorless GoTaq™ flexi Buffer, 2 x 1mL 5X green GoTaq™ flexi Buffer. Store at −30 to −10°C
200 Reactions
Standard PCR, RT-PCR
200