missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GSH-Glo™ Glutathione Assay
The GSH-Glo™ Assay is a luminescent-based assay for the detection and quantification of glutathione (GSH) in cells or in various biological samples.
$926.27 - $3,865.84
Specifications
| Content And Storage | -30°C to -10°C |
|---|---|
| For Use With (Application) | For detection and quantification of Glutathione in cells or in various biological samples |
Includes: Reconstitution Buffer with esterase, Luciferin-NT, GSH-Glo™ Reaction Buffer, Glutathione S-Transferase, Glutathione (5mM), Luciferin Detection Reagent
Description
- A Luminescent-Based Assay for the Detection and Quantification of Glutathione (GSH)
- Measure reduced and/or total glutathione levels in cells and tissues
- Measure GSH levels directly from cell culture wells
- Excellent sensitivity and easily scalable
Specifications
| -30°C to -10°C | |
| For detection and quantification of Glutathione in cells or in various biological samples |