Promega HaloTag™ Vectors
A variety vectors for mammalian, E. coli, and cell-free protein expression
Supplier: Promega G9651
Description
- Appends HaloTag(R) to C-Terminus of Protein Fusion Partner
- Enables constitutive protein expression in mammalian cells
- Includes CMV immediate-early enhancer/promoter
- Suited to stable or transient gene expression
- Patent pending.
- For research use only. Researchers may use this product in their own research and they may transfer derivatives to others for research use provided that at the time of transfer a copy of this label license is given to the recipients and recipients agree to be bound by the terms of this label license. Researchers shall have no right to modify or otherwise create variations of the nucleotide sequence of the HaloTag gene. Researchers may however clone heterologous DNA sequences at either or both ends of said HaloTag gene so as to create fused gene sequences provided that the coding sequence of the resulting HaloTag gene has no more than four (4) deoxynucleotides missing at the affected terminus when compared to the intact HaloTag gene sequence. In addition, researchers must do one of the following: (1) use Promega HaloTag ligands, which can be modified or linked to Promega or customer-supplied moieties, or (2) contact Promega to obtain a license. No other use or transfer of this product or its derivatives is authorized without the express, written consent of Promega including, without limitation, Commercial Use. Commercial Use means any and all uses of this product and derivatives by a party for monetary or other consideration and may include but is not limited to use in: (1) product manufacture; and (2) to provide a service, information or data; and/or resale of the product or its derivatives, whether or not such product or derivatives are resold for use in research. With respect to such Commercial Use, or any diagnostic, therapeutic or prophylactic uses, please contact Promega for supply and licensing information. If the purchaser is not willing to accept the conditions of this limited use statement, Promega is willing to accept return of the unopened product and provide purchaser with a full refund. However, in the event that the product is opened, then purchaser agrees to be bound by the conditions of this limited use statement.
- For research use only. Persons wishing to use this product or its derivatives in other fields of use, including without limitation, commercial sale, diagnostics or therapeutics, should contact Promega Corporation for licensing information.
- Mfr. Nos. G2841, G2851, G2861, G2871, G2881, G2981, G3780, G2751, G2681, G1891, G1841, G1681, G1691: Licensed for research and laboratory use only under U.S. Patent 5,547,862.
Specifications
CMV | |
10mM Tris HCl (pH 8), 1mM EDTA | |
100 ng/μL | |
pFC14A HaloTag™ CMV Flexi™ vector |
High | |
-30°C to -10°C | |
20 μg | |
Cell imaging of Protein localization or trafficking in conjunction with the fluorescent HaloTag™ ligands by using expression of HaloTag™ fusion Proteins in E. coli and cell-free Protein expression |