Promega Luciferase Assay Systems DFS Item
An extremely sensitive reagent for rapid quantitation of firefly luciferase. Can be used for assays in single-tube luminometers or in multiwell-plate luminometers with auto-injectors.
Supplier:  Promega E4530
 View more versions of this product
Catalog No. PR-E4530
Explore available promotions
Description
Description
  • Rapid Detection of Firefly Luciferase Activity
  • Linear over eight or more orders of magnitude of enzyme concentration.
  • Sensitive to 10-20 moles of luciferase.
  • Cell lysis, sample prep and assay complete in as little as 5 minutes
  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  • Certain applications of this product may require licenses from others.
Specifications
Specifications
Luciferase Assay System Freezer Pack
4.6
1000 Assays
10 × 1 vial Luciferase Assay Substrate, 10 × 10mL Luciferase Assay Buffer, 10 × 30mL Reporter Lysis 5X Buffer
-30°C to -10°C
Liquid