Promega Magne™ Protein G Magnetic Affinity Beads DFS Item
Magne Protein G & Magne Protein A Beads are magnetic affinity beads with high specificity and capacity for purification of immunoglobulins from cell culture media, ascites and serum. Iron encapsulated in macroporous cellulose with low nonspecific binding.
Supplier:  Promega G7471
 View more versions of this product
Catalog No. PRG7471
Explore available promotions
Description
Description
  • High Specificity, High Capacity Magnetic Affinity Beads for Antibody Capture
  • Binding capacities of >25mg/mL depending on antibody species, isotype
  • Minimize purification losses and increase sample throughput with exceptional magnetic properties
Specifications
Specifications
≤25mg/mL
2°C to 10°C
1 mL
Magnetic Affinity Bead
20% Slurry in 20% Ethanol
30 to 80 μm
Antibody purification from Serum, ascites and cell-media samples, auto mated, high-throughput Antibody purification, Antibody purification and concentration from small-volume or dilute source material