missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Mass Spec Grade Trypsin/Lys-C Mix
Trypsin/Lys-C Mix, Mass Spec Grade, is a mixture of Trypsin Gold, Mass Spectrometry Grade, and rLys-C, Mass Spec Grade. The Trypsin/Lys-C Mix is designed to improve digestion of proteins or protein mixtures in solution.
$113.19 - $294.83
Specifications
Color | Colorless |
---|---|
For Use With (Application) | In-solution Protein digestion, in-solution digestion of proteolytically resistant Proteins |
Storage Buffer | 50mM Acetic Acid (pH <3) Resuspension Buffer |
Content And Storage | -30°C to -10°C |
Product Type | Trypsin/Lys-C Mix |
Catalog Number | Mfr. No. | Components | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Components | Quantity | Price | Quantity | ||||
PRV5071
|
Promega
V5071 |
20μg Mass spec grade lyophilized Trypsin/Lys-C Mix, 500μL Resuspension Buffer | 20 μg |
Each for $113.19
|
|
||||
PRV5072
|
Promega
V5072 |
100μg Mass spec grade lyophilized Trypsin/Lys-C Mix, 500μL Resuspension Buffer | 100 μg |
Each for $280.35
|
|
||||
PRV5073
|
Promega
V5073 |
5 x 20μg Mass spec grade lyophilized Trypsin/Lys-C Mix, 500μL Resuspension Buffer | 100 μg |
Each for $294.83
|
|
||||
Description
- Improve Protein Digestion with this Mix of Trypsin Gold and rLys-C
- Use standard overnight digestion, nondenaturing conditions
- Increased peptide recovery, enhanced protein quantitation, improved reproducibility
- Tolerant to trypsin-inhibiting contaminants
Specifications
Colorless | |
50mM Acetic Acid (pH <3) Resuspension Buffer | |
Trypsin/Lys-C Mix |
In-solution Protein digestion, in-solution digestion of proteolytically resistant Proteins | |
-30°C to -10°C | |
Liquid |