Promega ONE-Glo™ Assay
The ONE-Glo™ + Tox Assay combines luciferase assay chemistry with a cell viability marker to better understand reporter gene expression in the context of cell health. The assay uses a two-step, addition-only process to measure in a single well of a plate.
$496.73 - $3079.23
Description
- Sensitive Detection of Reporter Activity and Cell Health
- Measures cell viability and firefly luciferase activity in the same assay well
- Easy to use—simply add reagent, mix and read
- Scalable to meet throughput needs, up to 1,536-well format
Specifications
-30°C to -10°C | |
7.5 | |
Fluid |