Promega PDE-Glo™ Phosphodiesterase Assay DFS Item
The PDE-Glo™ Phosphodiesterase Assay is a luminescent, high-throughput screening method for measuring cyclic nucleotide phosphodiesterase activity from purified sources.
Supplier:  Promega V1361
Catalog No. PRV1361
Explore available promotions
Description
Description
  • Detects Cyclic Nucleotide Phosphodiesterase Activity
  • Assay complete in >1 hour after PDE reaction
  • Scale from 1-tube to 1536-well plate formats
  • Simple luminescence detection
Specifications
Specifications
1 x 1.2mL 5X PDE-Glo™ Reaction Buffer, 1 x 600μL 5X PDE-Glo™ detection Buffer, 1 x 20μL Protein Kinase A, 1 x 600μL 5X PDEGlo Termination Buffer, 1 x 500μL
-30°C to -10°C
Fluid
7.5
1000 Assays
Videos
SDS
Documents
Documents
Promotions
Promotions
  • Patent pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.