missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega pGL4 Luciferase Reporter Vectors DFS Item

Available in numerous configurations for enhanced usability and convenience

$708.87 - $785.29

Specifications

Content And Storage -30°C to -10°C
Concentration 1 μg/μL
Quantity 20 μg
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
View More Specs

Products 34
Catalog Number Mfr. No. Reporter Gene Storage Buffer Vector Price Quantity  
Catalog Number Mfr. No. Reporter Gene Storage Buffer Vector Price Quantity  
PRE6651 DFS Item
View Documents
Promega
E6651
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.10[luc2] vector
Each for $785.29
 
PRE6661 DFS Item
View Documents
Promega
E6661
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.11[luc2P] vector
Each for $783.97
 
PRE6681 DFS Item
View Documents
Promega
E6681
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.13[luc2/SV40] vector
Each for $783.97
 
PRE6721 DFS Item
View Documents
Promega
E6721
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.17[luc2/Neo] vector
Each for $783.97
 
PRE6731 DFS Item
View Documents
Promega
E6731
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.18[luc2P/Neo] vector
Each for $783.97
 
PRE6911 DFS Item
View Documents
Promega
E6911
hRlucB 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.73[hRluc/SV40] vector
Each for $783.97
 
PRE6771 DFS Item
View Documents
Promega
E6771
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.22[luc2CP/Puro] vector
Each for $783.97
 
PRE6881 DFS Item
View Documents
Promega
E6881
hRlucB 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.70[hRluc] vector
Each for $783.97
 
PRE6921 DFS Item
View Documents
Promega
E6921
hRlucB 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.74[hRluc/TK] vector
Each for $783.97
 
PRE6931 DFS Item
View Documents
Promega
E6931
hRlucB 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.75[hRluc/CMV] vector
Each for $783.97
 
PR-E6751 DFS Item
View Documents
Promega
E6751
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.20[luc2/Puro] vector
Each for $783.97
 
PRE6701 DFS Item
View Documents
Promega
E6701
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.15[luc2P/Hygro] vector
Each for $783.97
 
PR-E6691 DFS Item
View Documents
Promega
E6691
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.14[luc2/Hygro] vector
Each for $783.97
 
PR-E8491 DFS Item
View Documents
Promega
E8491
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.32[luc2P/NF-κB-RE/Hygro] vector
Each for $783.97
 
PRE1340 DFS Item
View Documents
Promega
E1340
luc2A 10mM Tris HCl, 1mM EDTA (pH 8) pGL4.33[luc2P/SRE/Hygro] vector
Each for $783.97
 
PRE1350 DFS Item
View Documents
Promega
E1350
luc2A 10mM Tris HCl, 1mM EDTA (pH 8) pGL4.34[luc2P/SRF-RE/Hygro] vector
Each for $783.97
 
PRE1360 DFS Item
View Documents
Promega
E1360
luc2A 10mM Tris HCl, 1mM EDTA (pH 8) pGL4.36[luc2P/MMTV/Hygro] vector
Each for $783.97
 
PRE1370 DFS Item
View Documents
Promega
E1370
luc2A 10mM Tris HCl, 1mM EDTA (pH 7.4 at 25°C) pGL4.35[luc2P/9XGAL4UAS/Hygro] vector
Each for $783.97
 
P3935A DFS Item
View Documents
Promega
C9351
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.31[luc2P/GAL4UAS/Hygro] vector
Each for $735.23
 
PRE1310 DFS Item
missing translation for 'viewDocuments'
Promega
E1310
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.50[ luc2 /CMV/Hygro] vector
Each for $708.87
 
PRE1320 DFS Item
missing translation for 'viewDocuments'
Promega
E1320
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.51[luc2/CMV/Neo] vector
Each for $708.87
 
PRE8411 DFS Item
missing translation for 'viewDocuments'
Promega
E8411
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.23[luc2/minP] vector
Each for $783.97
 
PRE8431 DFS Item
missing translation for 'viewDocuments'
Promega
E8431
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.25[luc2CP/minP] vector
Each for $783.97
 
PRE8441 DFS Item
missing translation for 'viewDocuments'
Promega
E8441
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.26[luc2/minP/Hygro] vector
Each for $783.97
 
PRE8421 DFS Item
View Documents
Promega
E8421
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.24[luc2P/minP] vector
Each for $783.97
 
PRE8451 DFS Item
View Documents
Promega
E8451
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.27[luc2P/minP/Hygro] vector
Each for $783.97
 
PRE8481 DFS Item
View Documents
Promega
E8481
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.30[luc2P/NFAT-RE/Hygro] vector
Each for $783.97
 
PRE8461 DFS Item
View Documents
Promega
E8461
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.28[luc2CP/minP/Hygro] vector
Each for $783.97
 
PRE8471 DFS Item
View Documents
Promega
E8471
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.29[luc2P/CRE/Hygro] vector
Each for $783.97
 
PRE3641 DFS Item
View Documents
Promega
E3641
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.37[luc2P/ARE/Hygro] vector
Each for $783.97
 
PRE4001 DFS Item
View Documents
Promega
E4001
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.42[luc2P/HRE/Hygro] vector
Each for $783.97
 
PRE4121 DFS Item
View Documents
Promega
E4121
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.43[luc2P/XRE/Hygro] vector
Each for $783.97
 
PRE4111 DFS Item
View Documents
Promega
E4111
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.44[luc2P/AP1 RE/Hygro] vector
Each for $783.97
 
PRE4651 DFS Item
View Documents
Promega
E4651
luc2A 10mM Tris HCl (pH 7.4), 1mM EDTA pGL4.52[luc2P/STAT5RE/Hygro] vector
Each for $783.97
 
Description

Description

  • Configurations available with the synthetic firefly luc2 (Photinus pyralis) and Renilla hRluc (Renilla reniformis) genes, which have been codon optimized for more efficient expression in mammalian cells
  • Both the reporter genes and the vector backbone, including the bla (β-lactamase or Ampr) and mammalian selectable marker genes for hygromycin (Hygro or Hygr), neomycin (Neo or Neor) and puromycin (Puro or Puror), have been engineered to reduce the number of consensus transcription factor binding sites, reducing background and the risk of anomalous transcription
  • The backbone is also supplied with two Rapid Response™ Luciferase Reporter genes (-P, -CP) for each luciferase gene; the proteins encoded by these Rapid Response genes respond more quickly and with greater magnitude to changes in transcriptional activity than their more stable counterparts
  • Choice of mammalian selectable markers provides easy transition from transient to stable cells
  • Use of a common multiple cloning site and a unique Sfil transfer scheme permits easy transfer from vector to vector

Vector Options

  • Basic vectors with no promoter that contain a multiple cloning region for cloning a promoter of choice
  • Vectors containing a minimal promoter
  • Vectors containing response elements (cAMP, NFAT, NF-κB, GAL4UAS) and a minimal promoter
  • Promoter-containing vectors that can be used as expression controls or as coreporter vectors

Transcription regulation, Virus-cell interactions, Compound screening, Post-translational modifications, GPCR signaling, Cell signaling, Promoter analysis

  • For research use only. This product and/or its use is subject to one or more of the following Promega patents: U.S. Pat. Appln. Ser. Nos. 09/645,706; 10/664,341; 10/943,508; U.S. Patents 5,670,356; 6,387,675 and 6,552,179, Australian Patent 698424 and various corresponding patent applications and issued patents. The terms of the limited license conveyed with the purchase of this product are as follows: Researchers may use this product in their own research and they may transfer derivatives to others for such research use provided that at the time of transfer a copy of this label license is given to the recipients and the recipients agree to be bound by the conditions of this label license. Researchers shall have no right to modify or otherwise create variations of the nucleotide sequence of the luciferase gene except that Researchers may: (1) clone heterologous DNA sequences at either or both ends of said luciferase gene so as to create fused gene sequences provided that the coding sequence of the resulting luciferase gene has no more than four deoxynucleotides missing at the affected terminus when compared to the intact luciferase gene sequence, and (2) insert and remove nucleic acid sequences in furtherance of splicing research predicated on the inactivation or reconstitution of the luminescent activity of the encoded luciferase. In addition, Researchers must do one of the following: (1) use luminescent assay reagents purchased from Promega Corporation for all determinations of luminescence activity resulting from the research use of this product and its derivatives; or (2) contact Promega to obtain a license for the use of the product and its derivatives. No other use or transfer of this product or its derivatives is authorized without the express written consent of Promega including, without limitation, Commercial Use. Commercial Use means any and all uses of this product and derivatives by a party for monetary or other consideration and may include but is not limited to use in: (1) product manufacture; and (2) to provide a service, information or data; and/or resale of the product or its derivatives, whether or not such product or derivatives are resold for use in research. With respect to such Commercial Use, or any diagnostic, therapeutic or prophylactic uses, please contact Promega for supply and licensing information. If the purchaser is not willing to accept the conditions of this limited use statement, Promega is willing to accept the return of the unopened product and provide the purchaser with a full refund. However, in the event the product is opened, then the purchaser agrees to be bound by the conditions of this limited use statement.
  • Mfr. No. C9351, E1310, E1320, E1370, E6651, E6661, E6671, E6681, E6691, E6701, E6711, E6721, E6731, E6741, E6751, E6761, E6771, E8411, E8421, E8431, E8441, E8451, E8461, E8471, E8481, E8491: The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
  • Mfr. No. E6881, E6891, E6901, E6911, E6921, E6931, E6941, E6951, E6961, E6971, E6981, E6991, E7501, E7511, E7521: Licensed from University of Georgia Research Foundation, Inc., under U.S. Patent 5,418,155; Canadian Patents 2,105,984 and related patents.
  • Mfr. No. E8481: The NFAT response element and its use are licensed under one or more of the following patents: U.S. Patents 5,837,840; 6,197,925; 5,989,810; 6,096,515; 6,388,052; 6,150,099; 6,171,781; 6,352,830; 6,312,899 and 6,875,571.
  • Mfr. No. E1310, E1320: The use of this product and derivatives thereof is strictly limited to that of a life sciences research reagent. All other use is strictly prohibited, including but not limited to any diagnostic, th

Specifications

Specifications

-30°C to -10°C
1 μg/μL
20 μg
Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Videos
SDS
Documents

Documents