Learn More
Promega Proteasome-Glo™ Cell-Based Assays DFS Item
The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells.
Supplier:  Promega G8760
 View more versions of this product
Catalog No. PR-G8760
Only null left
Explore available promotions
Description
Description
  • Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
  • Simple add-mix-measure protocol
  • Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
  • Results within 10-30 minutes after reagent addition
Specifications
Specifications
Trypsin-like Cell-based Assay
Bioluminescence
-30°C to -10°C
10 mL
1 x 15μL Inhibitor 1, 1 x 100μL Inhibitor Mix 2, 1 x 100μL Z LRR Glo substrate, 1 x 10mL Proteasome Glo Cell based Buffer, 1 x 1 each Luciferin detection Reagent
7.2
Proteasome-Glo
Liquid
Product Suggestions
Product Suggestions