Learn More
Promega Proteasome-Glo™ Cell-Based Assays
The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells.
Supplier: Promega G8660
Description
- Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
- Simple add-mix-measure protocol
- Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
- Results within 10-30 minutes after reagent addition
- U.S. Pat. No. 6,602,677, Australian Pat. No. 754312 and other patents and patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
| Chymotrypsin-like Cell-based Assay | |
| 7.2 | |
| 10 mL |
| 1 x 50μL Suc LLVY Glo substrate, 1 x 10mL Proteasome Glo Cell based Buffer, 1 x 1 each Luciferin detection Reagent | |
| -30°C to -10°C | |
| Liquid |