missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Proteinase K
Serine protease that exhibits a very broad cleavage specificity. Useful in many molecular biology applications.
Supplier: Promega V3021
Description
- For General Digestion of Protein in Biological Samples
- Active over a pH range of 4.3-12.0, in 0.5% SDS or 1% Triton(R) X-100
- Purified to remove RNase and DNase activities
Specifications
| 50 to 100 μg/mL | |
| 4.3 to 12 | |
| 50mM Tris HCl (pH 8), 10mM CaCl2 | |
| Tritirachium album limber | |
| Proteinase K | |
| PMSF (1) or DFP |
| Colorless | |
| Cleaves Peptide bonds adjacent to the Carboxylic group of Aliphatic and Aromatic Amino Acids, useful for general digestion of Protein in biological samples | |
| -30°C to -10°C | |
| 100 mg | |
| 28900 | |
| Lyophilized Powder |