Promega pTARGET™ Mammalian Expression Vector System
Convenient system for cloning and expression of PCR products in mammalian cells.
Supplier: Promega A1410
Includes: Vector; Control insert DNA; T4 DNA Ligase 10X Buffer; T4 DNA Ligase; High-efficiency competent cells
Description
- Simple PCR Cloning for Mammalian Expression
- Clone PCR products with T overhangs
- CMV promoter for robust, consistent expression
- Neomycin marker for selection of stable transfectants
Specifications
CMV | |
20 Reactions | |
Cloning PCR products and for expressing cloned PCR products in Mammalian cells |
<-65°C | |
pTargeT vector |