Promega ReliaPrep™ gDNA Tissue Miniprep System
The ReliaPrep gDNA Tissue Miniprep System provides a complete, ready-to-use method for purification of gDNA from up to 25mg of tissue, a buccal swab, or a 1cm mouse tail snip, obtaining intact gDNA without the use of ethanol washes or precipitations.
Supplier: Promega A2051
Description
- Manual DNA Extraction from 25mg Tissue, 1 Buccal Swab, or 1cm Mouse Tail
- No ethanol, eliminating alcohol inhibition and carryover.
- Good recovery and purity in as little as 50μL elution
- Consistent results from run-to-run and user-to-user
Specifications
15°C to 30°C | |
Purifies genomic DNA from up to 25mg of tissue, a buccal swab or a 1cm Mouse tail snip, obtaining intact genomic DNA without the use of Ethanol washes or precipitations | |
1 x 50mL Binding Buffer, 1 x 165mL Column wash solution, 2 x 50 ReliaPrep™ binding columns, 2 x 1.1mL Proteinase K solution, 1 x 40mL Cell Lysis Buffer, 10 x 40 collection tubes, 1 x 10mL tail Lysis Buffer, 1 x 250μL RN | |
100 Preps |