Learn More
Promega RQ1 RNase-Free DNase
RQ1 (RNA Qualified) RNase-Free DNase is a DNase I that degrades both double-stranded and single-stranded DNA.
Supplier: Promega M6101
Description
- Degrades Single-Stranded or Double-Stranded DNA
- Produces 3′ hydroxyl oligonucleotides during degradation
- Preparation qualified for use in applications where maintaining the integrity of RNA is critical
- Supplied at a concentration of 1u/μL
Specifications
| 1 U/μL | |
| 7.5 | |
| 20mM HEPES (7.5 pH), 10mM CaCl2, 10mM MgCl2, 50% Glycerol | |
| Bovine pancreas | |
| RQ1 RNase-free DNase |
| RQ1 DNase 10X Reaction Buffer 1 x 1mL, RQ1 DNase Stop Solution 1 x 1mL, RQ1 RNase-Free Dnase 1 x 1000U | |
| Preparation of DNA-free RNA, degradation of DNA from RNA transcription systems, nick translation of DNA, studying DNA : Protein interactions by DNase I footprinting | |
| -30°C to -10°C | |
| 1000 U | |
| Liquid |