Learn More
Promega Vac-Man™ Vacuum Manifold
For use with any Wizard™ DNA Purification System or SV Total RNA Isolation system
Supplier: Promega A7231
Description
- Processes up to 20 samples simultaneously, while allowing individual control of each vacuum port
- Eliminates the need to handle waste during nucleic acid purification
- Includes Manifold, Stopcocks, Neoprene Stopper and Spinlock II Adapters
Specifications
| 15°C to 30°C | |
| Wizard™ Nucleic Acid purification products | |
| Simple and reliable method for rapid isolation of plasmid DNA | |
| 1/Ea. |