Promega Wizard™ Genomic DNA Purification Kits DFS Item
An easy, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.
Supplier:  Promega A1120
 View more versions of this product
Catalog No. PR-A1120
Explore available promotions
Includes: Cell lysis solution, nuclei lysis solution, protein precipitation solution, DNA rehydration solution, RNase solution‡
Description
Description
  • Manual, Solution-Based DNA Extraction Method
  • DNA from blood, animal and plant cells, bacteria or yeast
  • 60-minute, scalable protocol
  • Kit size based on number of isolations from whole blood, other sample types may vary
† To fully rehydrate DNA, allow an additional hour at 65°C (149°F) or overnight at room temperature.
‡ PR-A1620 does not include RNase Solution.
Specifications
Specifications
15°C to 30°C
100mL Cell lysis solution, 50mL Nuclei lysis solution, 25mL Protein precipitation solution, 50mL DNA rehydration solution, 250μL RNase solution
Genomic DNA Purification Kit
5 to 15μg/300μL
Isolate genomic DNA from whole blood, cultured cells, yeast, mouse tissue or numerous other cell sources
100 Preps
300μL